Learn More
Pfanstiehl Inc a. a-Trehalose Dihydrate (Inhalation Grade) High Purity, Low Endotoxin, Low Metals, USP-NF, EP, JP, ChP (T-104-4-IG) 1KG, GMP , Multi-compendial CAS: 6138-23-4 Injectable Excipient Grade
Supplier:  Pfanstiehl Inc T1044IG1KG
Encompass
Pfanstiehl Trehalose - High Purity, Low Endotoxin, Low Metals is a multi-compendial stabilizer of proteins, lipids, and carbohydrates in biologic therapeutics. The Inhalation Grade Trehalose was developed and tested for pharmaceutical Inhalation formulations. Used throughout formulation and freeze/thaw cycles, Trehalose ensures stability and efficacy in biopharmaceutical applications. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Trehalose meets parenteral (injectable) excipient grade standards, making it ideal for biologic and vaccine formulations. It is also widely used in cryopreservation media to protect cell membranes and enhance cell recovery for bioproduction and cell therapy. Pfanstiehl Brand Trehalose is multi-compendial and available as USP-NF (National Formulary), EP (Ph. Eur.) (European Pharmacopoeia), JP (Japanese Pharmacopoeia), and ChP (Chinese Pharmacopoeia).
Catalog No. NC2063109