Learn More
Medchemexpress LLC HY-19610 1mg , α-Amanitin CAS:23109-05-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY196101MG
Encompass_Preferred
HY-19610 1mg α-Amanitin CAS: 23109-05-9 α-Amanitin is the principal toxin of several deadly poisonous mushrooms, exerting its toxic function by inhibiting RNA-polymerase II. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9385