Learn More
ATCC ATCC Quantitative Genomic RNA, Influenza A (H3N2) strain A/Baltimore/JH-335/2022 (VR-3398D) - 100 μL, frozen
Supplier:  ATCC VR3398D
Encompass
Quantitative Genomic RNA from Influenza A virus (H3N2) strain A/Baltimore/JH-335/2022 is a prepared nucleic acid standard supplied in a quantitative format for use in molecular assays such as quantitative PCR (qPCR). It is intended for assay development, verification, and validation in research settings and supports generation of standard curves to determine viral load.
  • Prepared quantitative RNA standard for qPCR.
  • Suitable for assay development, verification, and validation.
  • Enables standard curve generation for viral load determination.
  • Supplied frozen in a 100 μL volume.
  • Requires storage at -70°C or colder.
  • For laboratory research use only; not for diagnostic or therapeutic use.
Catalog No. NC3760735