Learn More
Cayman Chemical a-BoswelIc AcId 10mg
SDP
Supplier:  Cayman Chemical 1169210 MG
Encompass_Preferred
A pentacyclic triterpene; increases Survivin and Reelin expression, decreases production of reactive oxygen species (ROS) and levels of hyperphosphorylated Tau, and inhibits apoptosis induced by streptozotocin in primary fetal human astrocytes; increases gastric juice pH and production of gastric wall mucus and decreases leukocyte infiltration, submucosal edema, and the formation of hemorrhagic lesions in a rat model of ethanol-induced gastric injury at 200 mg/kg
Catalog No. 50-295-1731