Learn More
Bioworld A.C.E. Buffer TINTED (10X), 1 L
Supplier:  Bioworld 214200042
Encompass_Preferred
10X running buffer concentrate.Tinted light pink for easy differentiation from other solvents in the sequencing apparatus.
Catalog No. 50-198-613