Learn More
Bioworld ACE Phosphate Buffer, 50mM, pH 7.0, 250 mL
Supplier:  Bioworld 214200102
Encompass_Preferred
Product DescriptionA.C.E. TM phosphate buffer has a concentration of 50 mM with a pH level of 7.0. Phosphate Buffers have been specially formulated for capillary electrophoresis. They are ideal for the separation of various biomolecules including proteins. A.C.E. TM buffers deliver consistent and reproducible results because they are manufactured with the highest quality reagents in nuclease free water. Product is shipped at room temperature.
Catalog No. 50-198-625