Learn More
Bioworld A.C.E. Sequencing Buffer Concentrate (10X), 1 L
Supplier:  Bioworld 214200001
Encompass_Preferred
Conductivity @25C(mS)- 5.78-6.72 DNase(P/F) - None pH @25C - 7.60-7.80 RNase(P/F) - None
Catalog No. 50-198-604