Learn More
Cayman Chemical a-CembrendIol 10mg
SDP
Supplier:  Cayman Chemical 3604210 MG
Encompass_Preferred
A diterpenoid with diverse biological activities; inhibits induction of EA-EBV by TPA in Raji cells (IC50 = 25.2 μM); active against the plant pathogenic fungus V. mali (IC50 = 18 mg/L); dermal application (3.3 µmol) reduces the tumor promoting effects of TPA on 7,12-demethylbenz[a]anthracene-induced papilloma formation by 53% in mice; has been found in cigarette smoke condensate
Catalog No. 50-291-3032