Learn More
Bachem Human alpha-CGRP Trifluoroacetate | 90954-53-3 | MFCD00285485 | 1mg
SDP
Supplier:  Bachem 4013281.1000
Encompass_Preferred
Human alpha-CGRP Trifluoroacetate | 90954-53-3 | MFCD00285485 | 1mg. Stored at <-15°C and shipped at ambient temperature.
Catalog No. NC9074088