Learn More
Cayman Chemical a-CyclodextrInhydrate 10g
SDP
Supplier:  Cayman Chemical 3121910 G
Encompass_Preferred
A cyclic hexasaccharide; forms inclusion complexes with wide variety of molecules, improving their solubility and stabilization; has commonly been used in the separation and purification of chemicals on an industrial scale; has also been used in the pharmaceutical and food industries
Catalog No. 50-299-0017