Learn More
Medchemexpress LLC a-D-Glucosamine pent 25g | 7784-54-5 | 25 G

Supplier: Medchemexpress LLC HY11499825G
α-D-Glucosamine pentaacetate (2-acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-α-D-glucopyranose) is an acetylated derivative of D-glucosamine used as a research reagent in glycobiology and related synthetic applications. The material is supplied as a solid with confirmed composition and batch-level quality testing.
- Acetylated derivative of D-glucosamine suitable for glycobiology research.
- Chemical name: 2-acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-α-D-glucopyranose.
- Molecular formula C16H23NO10; molecular weight 389.35.
- Purity ≥97.0% by NMR according to batch certificate of analysis.
- Supplied as a 25 G solid package.
- Intended for research, synthesis, and analytical applications (not for human use).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.