Learn More
STA PHARMACEUTICAL US LLC a-(Fmoc-amino)-3-tetrahydrofuran acetic acid | 1 g | CAS 1695476-98-2 | MDL MFCD28389395
Supplier: STA PHARMACEUTICAL US LLC STAAA101801GM
a-(Fmoc-amino)-3-tetrahydrofuran acetic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1695476-98-2
- MDL: MFCD28389395
- InChIKey: BLVWTMAIPKJGEW-UHFFFAOYSA-N
- Molecular Weight: 367.401
- Molecular Formula: C21H21NO5
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29321900
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-(tetrahydrofuran-3-yl)acetic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)NC(C4CCOC4)C(O)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.