Learn More
Enzo Life Sciences alpha Gal Epitope (Galalpha1-3Galbeta1-4GlcNAc-R) monoclonal antibody (M86) (1 Vial)
SDP
Supplier:  Enzo Life Sciences ALX8010901
Encompass_Preferred
Clone: M86. Host: Mouse. Isotype: IgM. Immunogen: Rabbit red blood cell membrane. Source: Hybridoma tissue culture. Species Reactivity: Mouse, Rat, Porcine. Specificity: Recognizes synthetic and naturally produced mouse, rat and pig Galα1-3Gal epitopes on glycoproteins and glycolipids. Crossreactivity: Does not cross-react with β-Gal glycoproteins or BSA. Applications: ELISA. Flow Cytometry, IHC (PS), WB. FUNC. Recommended dilutions: Note: Do not use non-fat milk for blocking! Many blocking agents from non-human mammal. Sources will cross-react with the antibody. Appropriate antibody dilution may vary from lot to lot. An initial dilution of 1:5 is recommended. Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Tissue culture supernatant. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. NC0277743