Learn More
Cayman Chemical (−)-(α)-Kainic Acid (hydrate), 58002-62-3, 50mg
SDP
Supplier:  Cayman Chemical 7805050
Encompass_Preferred
Molecular Formula C10H15NO4 • H2O, Purity ≥98%, Formula Weight 231.2, Synonyms: 2S-carboxy-4S-(1-methylethenyl)-3S-pyrrolidineacetic acid, monohydrate; A natural marine product originally isolated from the red marine alga D. simplex that is a potent CNS stimulant; developed as the prototype neuroexcitatory amino acid for the induction of seizures in experimental animals, at a typical dose of 10-30 mg/kg in mice; neuroexcitotoxic and epileptogenic, acting through specific kainate receptors.
Catalog No. NC0759764
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.