Learn More
Cayman Chemical a-KetoIsocaproIc AcIdodIum 5g
SDP
Supplier:  Cayman Chemical 210525 G
Encompass_Preferred
A metabolite of leucine catabolism; increases the production of reactive species and decreases the activity of mitochondrial complex I and complex II-III in the rat hippocampus at 4 µmol; urine levels are reduced in db/db diabetic mice compared with heterozygous db/m non-diabetic mice; accumulates in the tissues and body fluids of patients with maple syrup urine disease
Catalog No. 50-298-4175