Learn More
American Radiolabeled Chemicals Inc A-MSH NDP 125I 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARI 014250UC
Encompass
a-MSH, Nle4, D-Phe7-125I 50uCi in buffered solution (pH 8.5), 0.075 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1770, Shipped in Dry Ice
Catalog No. 50-901-021
Add to Cart
Request a Quote