Learn More
Indofine Chemical Α-Solanine With Hp, 98%, 10 Mg
SDP
Supplier:  Indofine Chemical 021652S 10 MG
Encompass_Preferred
-SOLANINE with HPLC 10 MG, Herbal Standards, Solatunine, C45H73NO15, MW: 868.08, >98% by HPLC, 20562-02-1, MFCD00077873
Catalog No. 50-909-054