Learn More
Cayman Chemical -a-Tocopherol 50g
SDP
Supplier:  Cayman Chemical 2598550 G
Encompass_Preferred
A biologically active form of vitamin E; inhibits brominated diphenyl ether-47-induced increases in ROS and PGE2 production in HTR-8/SVneo cells at 20 μM; inhibits lipid peroxidation in HepG2 cells (IC50 = 24.5 μM)
Catalog No. 50-293-3366