Learn More
Cayman Chemical a-TruxIlIc AcId 10mg
SDP
Supplier:  Cayman Chemical 1486510 MG
Encompass_Preferred
Can be formed by the dimerization of two molecules of α-trans-cinnamic acid; significantly blocks inflammatory pain while having little effect on neurogenic pain, as indicated by the formalin test in mice
Catalog No. 50-297-1357