Learn More
ABclonal Technology Tyrosine Hydroxylase Rabbit pAb
SDP
Supplier:  ABclonal Technology A0028500UL
Encompass_Preferred
The protein encoded by this gene is involved in the conversion of tyrosine to dopamine. It is the rate-limiting enzyme in the synthesis of catecholamines, hence plays a key role in the physiology of adrenergic neurons. Mutations in this gene have been associated with autosomal recessive Segawa syndrome. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Catalog No. 50-001-38368