Learn More
ABclonal Technology Integrin beta 3 (ITGB3/CD61) Rabbit pAb
SDP
Supplier:  ABclonal Technology A00761000UL
Encompass_Preferred
The ITGB3 protein product is the integrin beta chain beta 3. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. Integrin beta 3 is found along with the alpha IIb chain in platelets. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling.
Catalog No. 50-001-35693