Learn More
ABclonal Technology eEF2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A00991000UL
Encompass_Preferred
This gene encodes a member of the GTP-binding translation elongation factor family. This protein is an essential factor for protein synthesis. It promotes the GTP-dependent translocation of the nascent protein chain from the A-site to the P-site of the ribosome. This protein is completely inactivated by EF-2 kinase phosporylation.
Catalog No. 50-001-38804