Learn More
ABclonal Technology Hexokinase II Rabbit pAb
SDP
Supplier:  ABclonal Technology A0165500UL
Encompass_Preferred
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes hexokinase 2, the predominant form found in skeletal muscle. It localizes to the outer membrane of mitochondria. Expression of this gene is insulin-responsive, and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells.
Catalog No. 50-001-37940