Learn More
ABclonal Technology FABP4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A02321000UL
Encompass_Preferred
FABP4 encodes the fatty acid binding protein found in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
Catalog No. 50-001-37802