Learn More
ABclonal Technology IGF1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A03031000UL
Encompass_Preferred
The protein encoded by this gene is similar to insulin in function and structure and is a member of a family of proteins involved in mediating growth and development. The encoded protein is processed from a precursor, bound by a specific receptor, and secreted. Defects in this gene are a cause of insulin-like growth factor I deficiency. Alternative splicing results in multiple transcript variants encoding different isoforms that may undergo similar processing to generate mature protein.
Catalog No. 50-001-37283