Learn More
ABclonal Technology EEA1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A05921000UL
Encompass_Preferred
Enables 1-phosphatidylinositol binding activity, GTP-dependent protein binding activity, and protein homodimerization activity. Involved in endocytosis, vesicle fusion, and viral RNA genome replication. Located in cytosol and early endosome. Is extrinsic component of plasma membrane.
Catalog No. 50-001-36445