Learn More
ABclonal Technology MyoD1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A06711000UL
Encompass_Preferred
This gene encodes a nuclear protein that belongs to the basic helix-loop-helix family of transcription factors and the myogenic factors subfamily. It regulates muscle cell differentiation by inducing cell cycle arrest, a prerequisite for myogenic initiation. The protein is also involved in muscle regeneration. It activates its own transcription which may stabilize commitment to myogenesis.
Catalog No. 50-001-34901