Learn More
ABclonal Technology ATF2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A07571000UL
Encompass_Preferred
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. The encoded protein has been identified as a moonlighting protein based on its ability to perform mechanistically distinct functions This protein binds to the cAMP-responsive element (CRE), an octameric palindrome. It forms a homodimer or a heterodimer with c-Jun and stimulates CRE-dependent transcription. This protein is also a histone acetyltransferase (HAT) that specifically acetylates histones H2B and H4 in vitro, thus it may represent a class of sequence-specific factors that activate transcription by direct effects on chromatin components. The encoded protein may also be involved in cells DNA damage response independent of its role in transcriptional regulation. Several alternatively spliced transcript variants have been found for this gene
Catalog No. 50-001-35071