Learn More
ABclonal Technology LRRK2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A08591000UL
Encompass_Preferred
This gene is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat (LRR) domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane. Mutations in this gene have been associated with Parkinson disease-8.
Catalog No. 50-001-36036