Learn More
ABclonal Technology FABP5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A0947500UL
Encompass_Preferred
This gene encodes the fatty acid binding protein found in epidermal cells, and was first identified as being upregulated in psoriasis tissue. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs may play roles in fatty acid uptake, transport, and metabolism. Polymorphisms in this gene are associated with type 2 diabetes. The human genome contains many pseudogenes similar to this locus.
Catalog No. 50-001-35257