Learn More
ABclonal Technology PPP3R1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A0954500UL
Encompass_Preferred
Enables cyclosporin A binding activity, phosphatase binding activity, and protein domain specific binding activity. Involved in calcineurin-NFAT signaling cascade and positive regulation of transcription by RNA polymerase II. Part of calcineurin complex. Implicated in Alzheimers disease and dilated cardiomyopathy.
Catalog No. 50-001-35705