Learn More
New England Biolabs, Inc. α1-2,4,6 Fucosidase O - 400 units
SDP
Supplier:  New England Biolabs, Inc. P0749L
Encompass
a1-2,4,6 Fucosidase O is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal a1-2, a1-4 and a1-6 linked fucose residues from oligosaccharides. a1-2,4,6 Fucosidase O cleaves a1-6 fucose residues more efficiently than other linkages.
Catalog No. 50-152-6424