Learn More
Athens Research And Technology Inc A1 ANTITRYPSIN HU 1MG
Supplier:  Athens Research And Technology Inc 16160116091MG
Encompass_Preferred
Alpha 1 Antitrypsin, Human Plasma: 1 MG; Lyophilized from 30 mM Na phosphate, pH 6.5, with 300 mM NaCl; MW 52,000; Extinction Coefficient= 0.433; Purity: > =95% by SDS-PAGE; This native protein purified in the Athens Research & Tech lab.
Catalog No. 50-489-605