Learn More
Cayman Chemical A1-Phytoprostn-I 100UG
SDP
Supplier:  Cayman Chemical 9000593100 UG
Encompass_Preferred
A cyclopentenone isoprostane produced by the action of reactive oxygen species on α-linolenic acid in plants; induces the expression of glutathione-S-transferase, increases phytoalexin biosynthesis, and triggers the expression of several genes involved in primary and secondary metabolism in plants
Catalog No. 50-289-2599