Learn More
ABclonal Technology AP1M1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A101291000UL
Encompass_Preferred
The protein encoded by this gene is the medium chain of the trans-Golgi network clathrin-associated protein complex AP-1. The other components of this complex are beta-prime-adaptin, gamma-adaptin, and the small chain AP1S1. This complex is located at the Golgi vesicle and links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. Alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.
Catalog No. 50-001-35090