Learn More
ABclonal Technology RBM34 Rabbit pAb
SDP
Supplier:  ABclonal Technology A101391000UL
Encompass_Preferred
This gene encodes a member of the RNA-binding motif family of RNA recognition motif proteins. The encoded protein contains an RNA-binding domain made up of two RNA recognition motif subdomains referred to as RNA recognition motif-1 and RNA recognition motif-2. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-35093