Learn More
ABclonal Technology HRH4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10151500UL
Encompass_Preferred
Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Its various actions are mediated by a family of histamine receptors, which are a subset of the G-protein coupled receptor superfamily. This gene encodes a histamine receptor that is predominantly expressed in haematopoietic cells. The protein is thought to play a role in inflammation and allergy reponses. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-35818