Learn More
ABclonal Technology LY6E Rabbit pAb
SDP
Supplier:  ABclonal Technology A102251000UL
Encompass_Preferred
This gene belongs to the human Ly6 gene family and encodes a glycosylphosphatidyl-inositol (GPI)-anchored cell surface protein. The protein plays an important role in T cell physiology, oncogenesis and immunological regulation. The protein is also involved in modulation of viral infection by coronaviruses, SARS-CoV, MERS-CoV and SARS-CoV-2.
Catalog No. 50-001-38500