Learn More
ABclonal Technology EVPL Rabbit pAb
SDP
Supplier:  ABclonal Technology A102151000UL
Encompass_Preferred
This gene encodes a member of the plakin family of proteins that forms a component of desmosomes and the epidermal cornified envelope. This gene is located in the tylosis oesophageal cancer locus on chromosome 17q25, and its deletion is associated with both familial and sporadic forms of oesophageal squamous cell carcinoma. Patients suffering from the autoimmune mucocutaneous disorder, paraneoplastic pemphigus, develop antibodies against the encoded protein.
Catalog No. 50-001-38484