Learn More
ABclonal Technology 14-3-3 alpha/beta Rabbit pAb
SDP
Supplier:  ABclonal Technology A10231000UL
Encompass_Preferred
This gene encodes a protein belonging to the 14-3-3 family of proteins, members of which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals. The encoded protein has been shown to interact with RAF1 and CDC25 phosphatases, suggesting that it may play a role in linking mitogenic signaling and the cell cycle machinery. Two transcript variants, which encode the same protein, have been identified for this gene.
Catalog No. 50-001-35876