Learn More
ABclonal Technology DMT1/SLC11A2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10231500UL
Encompass_Preferred
This gene encodes a member of the solute carrier family 11 protein family. The product of this gene transports divalent metals and is involved in iron absorption. Mutations in this gene are associated with hypochromic microcytic anemia with iron overload. A related solute carrier family 11 protein gene is located on chromosome 2. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-38510