Learn More
ABclonal Technology EIF2B3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10262500UL
Encompass_Preferred
The protein encoded by this gene is one of the subunits of initiation factor eIF2B, which catalyzes the exchange of eukaryotic initiation factor 2-bound GDP for GTP. It has also been found to function as a cofactor of hepatitis C virus internal ribosome entry site-mediated translation. Mutations in this gene have been associated with leukodystrophy with vanishing white matter. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-36761