Learn More
ABclonal Technology DDAH1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A102951000UL
Encompass_Preferred
This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
Catalog No. 50-001-37542