Learn More
ABclonal Technology COG6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103191000UL
Encompass_Preferred
This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi apparatus. The encoded protein is organized with conserved oligomeric Golgi complex components 5, 7 and 8 into a sub-complex referred to as lobe B. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-38312