Learn More
ABclonal Technology LRRC4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103211000UL
Encompass_Preferred
Predicted to be involved in modulation of chemical synaptic transmission and synapse organization. Predicted to act upstream of or within synapse organization. Predicted to be located in neuron spine and postsynaptic membrane. Predicted to be active in Schaffer collateral - CA1 synapse, glutamatergic synapse, and plasma membrane. Predicted to be integral component of postsynaptic density membrane.
Catalog No. 50-001-34976