Learn More
ABclonal Technology CABLES1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10335500UL
Encompass_Preferred
This gene encodes a protein involved in regulation of the cell cycle through interactions with several cyclin-dependent kinases. One study (PMID 16177568) reported aberrant splicing of transcripts from this gene which results in removal of the cyclin binding domain only in human cancer cells, and reduction in gene expression was shown in colorectal cancers (PMID 17982127).Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-37517