Learn More
ABclonal Technology ZC3H14 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103641000UL
Encompass_Preferred
The protein encoded by this gene is a poly(A)-binding protein that can affect gene expression and poly(A) tail length. The encoded protein may influence mRNA stability, nuclear export, and translation.
Catalog No. 50-001-34973