Learn More
ABclonal Technology INPP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10372500UL
Encompass_Preferred
This gene encodes the enzyme inositol polyphosphate-1-phosphatase, one of the enzymes involved in phosphatidylinositol signaling pathways. This enzyme removes the phosphate group at position 1 of the inositol ring from the polyphosphates inositol 1,4-bisphosphate and inositol 1,3,4-trisphophosphate.
Catalog No. 50-001-37726