Learn More
ABclonal Technology CNDP2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103811000UL
Encompass_Preferred
CNDP2, also known as tissue carnosinase and peptidase A (EC 3.4.13.18), is a nonspecific dipeptidase rather than a selective carnosinase (Teufel et al., 2003 [PubMed 12473676]).
Catalog No. 50-001-35204