Learn More
ABclonal Technology COLEC12 Rabbit pAb
SDP
Supplier:  ABclonal Technology A104221000UL
Encompass_Preferred
This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. This protein is a scavenger receptor that displays several functions associated with host defense. It can bind to carbohydrate antigens on microorganisms, facilitating their recognition and removal. It also mediates the recognition, internalization, and degradation of oxidatively modified low density lipoprotein by vascular endothelial cells.
Catalog No. 50-001-35269